Product Details

SNP ID
rs143543321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36386183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCCTGGCACAACAAGGAGCCCG[C/T]GTACAGCCTGGACTTCCAGCATGGG
Phenotype
MIM: 601245 MIM: 610078
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHAF1B PubMed Links

Gene Details

Gene
CHAF1B
Gene Name
chromatin assembly factor 1 subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005441.2 221 Intron NP_005432.1
XM_011529755.2 221 UTR 5 XP_011528057.1
XM_017028477.1 221 Missense Mutation GCG,GTG A16V XP_016883966.1
XM_017028478.1 221 Missense Mutation GCG,GTG A16V XP_016883967.1
Gene
MORC3
Gene Name
MORC family CW-type zinc finger 3
There are no transcripts associated with this gene.

View Full Product Details