Product Details
- SNP ID
-
rs146427219
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:30166066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCTTTCTGTTGCAGCAGCAAAA[C/T]CCACAAAGCAGACCCCCTCCAATGA
- Phenotype
-
MIM: 617005
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN17
PubMed Links
Gene Details
- Gene
- CLDN17
- Gene Name
- claudin 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012131.2 |
588 |
Silent Mutation |
GGA,GGG |
G184G |
NP_036263.1 |
View Full Product Details