Product Details

SNP ID
rs146867345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32604135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAAACTACGTACCTGTGTTCCC[A/G]ACAAGTCTTCCTTATTTTCAAACTC
Phenotype
MIM: 615494
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C21orf59 PubMed Links

Gene Details

Gene
C21orf59
Gene Name
chromosome 21 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021254.2 889 Missense Mutation TCG,TTG S175L NP_067077.1

View Full Product Details