Product Details

SNP ID
rs149202971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30440734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCAGGAATTTCCAGGCAACTT[A/G]TTACTAACCAGCCTTTGGGTCTCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP13-4 PubMed Links

Gene Details

Gene
KRTAP13-4
Gene Name
keratin associated protein 13-4
There are no transcripts associated with this gene.

Gene
KRTAP15-1
Gene Name
keratin associated protein 15-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181623.1 407 Missense Mutation TAT,TGT Y136C NP_853654.1

View Full Product Details