Product Details

SNP ID
rs149483206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39725975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAACCTGAGCTCCAGCAACCTG[C/T]GGGTCCCAATGGAGGCGAAGTCAAG
Phenotype
MIM: 610638
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGSF5 PubMed Links

Gene Details

Gene
IGSF5
Gene Name
immunoglobulin superfamily member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080444.1 138 Intron NP_001073913.1
XM_011529472.1 138 UTR 5 XP_011527774.1
XM_011529473.1 138 UTR 5 XP_011527775.1
XM_017028279.1 138 UTR 5 XP_016883768.1
XM_017028280.1 138 UTR 5 XP_016883769.1

View Full Product Details