Product Details

SNP ID
rs138131052
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19482716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTATTTTATTCTCATAAACTG[C/T]GGAGCTAATGTAGACCTATTGGATA
Phenotype
MIM: 603465 MIM: 601754
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CDC45 PubMed Links

Gene Details

Gene
CDC45
Gene Name
cell division cycle 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178010.2 374 Silent Mutation TGC,TGT C77C NP_001171481.1
NM_001178011.2 374 Intron NP_001171482.1
NM_003504.4 374 Silent Mutation TGC,TGT C77C NP_003495.1
XM_005261285.2 374 Silent Mutation TGC,TGT C77C XP_005261342.1
XM_005261286.3 374 Silent Mutation TGC,TGT C77C XP_005261343.1
XM_011530415.1 374 Silent Mutation TGC,TGT C77C XP_011528717.1
XM_011530416.1 374 Silent Mutation TGC,TGT C65C XP_011528718.1
XM_011530417.2 374 Intron XP_011528719.1
XM_011530418.2 374 Intron XP_011528720.1
XM_017028966.1 374 Intron XP_016884455.1
XM_017028967.1 374 Intron XP_016884456.1
Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
There are no transcripts associated with this gene.

View Full Product Details