Product Details

SNP ID
rs138183274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23766207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACTCTGAGTGGTGGAACAGTC[C/T]AGGAACTGCCTGATCTCGTAGGCGC
Phenotype
MIM: 615903 MIM: 185261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182520.2 Intron NP_872326.2
XM_011529907.2 Intron XP_011528209.1
XM_011529908.2 Intron XP_011528210.2
XM_011529912.2 Intron XP_011528214.1
XM_017028602.1 Intron XP_016884091.1
XM_017028603.1 Intron XP_016884092.1
XM_017028604.1 Intron XP_016884093.1
XM_017028605.1 Intron XP_016884094.1
XM_017028606.1 Intron XP_016884095.1
XM_017028607.1 Intron XP_016884096.1
XM_017028608.1 Intron XP_016884097.1
XM_017028609.1 Intron XP_016884098.1
XM_017028610.1 Intron XP_016884099.1
XM_017028611.1 Intron XP_016884100.1
XM_017028612.1 Intron XP_016884101.1
XM_017028613.1 Intron XP_016884102.1
XM_017028614.1 Intron XP_016884103.1
XM_017028615.1 Intron XP_016884104.1
XM_017028616.1 Intron XP_016884105.1
XM_017028617.1 Intron XP_016884106.1
XM_017028618.1 Intron XP_016884107.1
Gene
CHCHD10
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301339.1 Intron NP_001288268.1
NM_213720.2 Intron NP_998885.1
Gene
MMP11
Gene Name
matrix metallopeptidase 11
There are no transcripts associated with this gene.

View Full Product Details