Product Details

SNP ID
rs141256918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26433753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGGATGGCCTGAGGGACTGCAT[C/T]GCCACCGGCATCCAGTCCGTGCGGG
Phenotype
MIM: 606682
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPHD2 PubMed Links

Gene Details

Gene
ASPHD2
Gene Name
aspartate beta-hydroxylase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020437.4 535 Silent Mutation ATC,ATT I46I NP_065170.2
Gene
HPS4
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
There are no transcripts associated with this gene.

View Full Product Details