Product Details

SNP ID
rs141606125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46583672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTTCTCATGGTGCGACTGGGG[A/C]CCTGGCTTAGCGGGAGGAAGACCAC
Phenotype
MIM: 613691
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GRAMD4 PubMed Links

Gene Details

Gene
GRAMD4
Gene Name
GRAM domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015124.4 Intron NP_055939.1
XM_005261396.3 Intron XP_005261453.1
XM_006724169.1 Intron XP_006724232.1
XM_006724170.3 Intron XP_006724233.1
XM_011530011.1 Intron XP_011528313.1
XM_011530012.1 Intron XP_011528314.1
XM_011530013.2 Intron XP_011528315.2
XM_011530014.2 Intron XP_011528316.1
XM_017028670.1 Intron XP_016884159.1

View Full Product Details