Product Details

SNP ID
rs141868904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41910000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCGGCATAGGGGCCCGGCGCG[A/G]CAGGGGGCGCGTAGACCGGCGGGGC
Phenotype
MIM: 612156 MIM: 600481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR33A PubMed Links

Gene Details

Gene
MIR33A
Gene Name
microRNA 33a
There are no transcripts associated with this gene.

Gene
SHISA8
Gene Name
shisa family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207020.1 1207 Missense Mutation GCC,GTC A320V NP_001193949.1
XM_006724254.3 1207 Missense Mutation GCC,GTC A415V XP_006724317.1
XM_006724255.2 1207 Missense Mutation GCC,GTC A379V XP_006724318.1
XM_006724256.3 1207 Missense Mutation GCC,GTC A375V XP_006724319.1
Gene
SREBF2
Gene Name
sterol regulatory element binding transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details