Product Details

SNP ID
rs142476770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20112708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGGATGAGCATCTCACAGTGC[A/G]GGGTCTGCGGGAACAGGTCCACTGC
Phenotype
MIM: 609030 MIM: 601180 MIM: 611151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190326.1 1983 Intron NP_001177255.1
NM_022720.6 1983 Intron NP_073557.3
XM_006724268.2 1983 Intron XP_006724331.1
Gene
MIR6816
Gene Name
microRNA 6816
There are no transcripts associated with this gene.

Gene
RANBP1
Gene Name
RAN binding protein 1
There are no transcripts associated with this gene.

Gene
TRMT2A
Gene Name
tRNA methyltransferase 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257994.1 1983 UTR 3 NP_001244923.1
NM_022727.5 1983 Missense Mutation CCG,CTG P578L NP_073564.3
NM_182984.4 1983 Missense Mutation CCG,CTG P578L NP_892029.2
XM_011530139.2 1983 Missense Mutation CCG,CTG P596L XP_011528441.1
XM_011530141.1 1983 Missense Mutation CCG,CTG P270L XP_011528443.1
XM_011530142.2 1983 Missense Mutation CCG,CTG P270L XP_011528444.1
XM_017028770.1 1983 Missense Mutation CCG,CTG P596L XP_016884259.1

View Full Product Details