Product Details

SNP ID
rs144666040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44755595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCGCGCTGCTCCGGAGTCCTGC[A/C]CCAGCTGCCTTCACAGGGCGTCTCC
Phenotype
MIM: 609405
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP8 PubMed Links

Gene Details

Gene
ARHGAP8
Gene Name
Rho GTPase activating protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017526.1 Intron NP_001017526.1
NM_001198726.1 Intron NP_001185655.1
NM_181335.2 Intron NP_851852.2
Gene
PRR5-ARHGAP8
Gene Name
PRR5-ARHGAP8 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181334.5 Intron NP_851851.3

View Full Product Details