Product Details

SNP ID
rs145238021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38426951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGGCCTGCGGCCACCGCGGC[C/T]GCCGCAGCTGCCTCCTCCTCCATCT
Phenotype
MIM: 600504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNJ4 PubMed Links

Gene Details

Gene
KCNJ4
Gene Name
potassium voltage-gated channel subfamily J member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004981.1 1279 Silent Mutation GCA,GCG A394A NP_004972.1
NM_152868.2 1279 Silent Mutation GCA,GCG A394A NP_690607.1
Gene
LOC101927183
Gene Name
uncharacterized LOC101927183
There are no transcripts associated with this gene.

View Full Product Details