Product Details

SNP ID
rs146551748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20112716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCATCTCACAGTGCGGGGTCTG[C/T]GGGAACAGGTCCACTGCCACAGCCT
Phenotype
MIM: 609030 MIM: 601180 MIM: 611151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190326.1 1975 Intron NP_001177255.1
NM_022720.6 1975 Intron NP_073557.3
XM_006724268.2 1975 Intron XP_006724331.1
Gene
MIR6816
Gene Name
microRNA 6816
There are no transcripts associated with this gene.

Gene
RANBP1
Gene Name
RAN binding protein 1
There are no transcripts associated with this gene.

Gene
TRMT2A
Gene Name
tRNA methyltransferase 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257994.1 1975 UTR 3 NP_001244923.1
NM_022727.5 1975 Silent Mutation CCA,CCG P575P NP_073564.3
NM_182984.4 1975 Silent Mutation CCA,CCG P575P NP_892029.2
XM_011530139.2 1975 Silent Mutation CCA,CCG P593P XP_011528441.1
XM_011530141.1 1975 Silent Mutation CCA,CCG P267P XP_011528443.1
XM_011530142.2 1975 Silent Mutation CCA,CCG P267P XP_011528444.1
XM_017028770.1 1975 Silent Mutation CCA,CCG P593P XP_016884259.1

View Full Product Details