Product Details
- SNP ID
-
rs146854087
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:40964640 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGGCCTTCCAAGAGTGTGACAGC[C/T]GTGAAGGATAACAAATATCAAATAT
- Phenotype
-
MIM: 603814
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBX1
PubMed Links
Gene Details
- Gene
- RBX1
- Gene Name
- ring-box 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014248.3 |
|
Intron |
|
|
NP_055063.1 |
View Full Product Details