Product Details

SNP ID
rs147415362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21030029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTAGGTGGTCTGAGAAGGAGCT[A/C]TGCTGCTTGGTCAGGGGGCCAGGGC
Phenotype
MIM: 608077 MIM: 603752
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR649 PubMed Links

Gene Details

Gene
MIR649
Gene Name
microRNA 649
There are no transcripts associated with this gene.

Gene
P2RX6
Gene Name
purinergic receptor P2X 6
There are no transcripts associated with this gene.

Gene
SLC7A4
Gene Name
solute carrier family 7 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004173.2 1373 Missense Mutation CAG,CAT Q435H NP_004164.2

View Full Product Details