Product Details

SNP ID
rs148631778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50486148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCATCCCCATCCTCGCTGGGCCC[A/G]CCGACCCCGGTGTTAGCAAGAATCC
Phenotype
MIM: 608682 MIM: 606774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADM2 PubMed Links

Gene Details

Gene
ADM2
Gene Name
adrenomedullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253845.1 3743 UTR 3 NP_001240774.1
XM_017028940.1 3743 UTR 3 XP_016884429.1
Gene
MIOX
Gene Name
myo-inositol oxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017584.5 3743 Intron NP_060054.4
XM_005261925.3 3743 Intron XP_005261982.1
XM_011530705.1 3743 UTR 5 XP_011529007.1

View Full Product Details