Product Details

SNP ID
rs148893857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37011180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGCGGTGACTCCCTTGCGGCA[C/T]GTGGCAATGAGAGGCTGCGAGAGAT
Phenotype
MIM: 602496 MIM: 180370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPST PubMed Links

Gene Details

Gene
MPST
Gene Name
mercaptopyruvate sulfurtransferase
There are no transcripts associated with this gene.

Gene
TEX33
Gene Name
testis expressed 33
There are no transcripts associated with this gene.

Gene
TST
Gene Name
thiosulfate sulfurtransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270483.1 784 Silent Mutation ACA,ACG T247T NP_001257412.1
NM_003312.5 784 Silent Mutation ACA,ACG T247T NP_003303.2

View Full Product Details