Product Details

SNP ID
rs150183795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30555556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGGGTCCAGGCTGTTGTCATA[G/T]CCCAGGTCGAAGAAGAGCAGGTTTC
Phenotype
MIM: 602300
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GAL3ST1 PubMed Links

Gene Details

Gene
GAL3ST1
Gene Name
galactose-3-O-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318103.1 843 Silent Mutation NP_001305032.1
NM_001318104.1 843 Silent Mutation NP_001305033.1
NM_001318105.1 843 Silent Mutation NP_001305034.1
NM_001318106.1 843 Silent Mutation NP_001305035.1
NM_001318107.1 843 Silent Mutation NP_001305036.1
NM_001318108.1 843 Silent Mutation NP_001305037.1
NM_001318109.1 843 Silent Mutation NP_001305038.1
NM_001318110.1 843 Silent Mutation NP_001305039.1
NM_001318111.1 843 Silent Mutation NP_001305040.1
NM_001318112.1 843 Silent Mutation NP_001305041.1
NM_001318113.1 843 Silent Mutation NP_001305042.1
NM_001318114.1 843 Silent Mutation NP_001305043.1
NM_001318115.1 843 Silent Mutation NP_001305044.1
NM_001318116.1 843 Silent Mutation NP_001305045.1
NM_004861.2 843 Silent Mutation NP_004852.1
XM_011530518.2 843 Silent Mutation XP_011528820.1
XM_011530522.2 843 Silent Mutation XP_011528824.1
XM_011530524.1 843 Silent Mutation XP_011528826.1
XM_011530528.1 843 Silent Mutation XP_011528830.1
XM_017029096.1 843 Silent Mutation XP_016884585.1
XM_017029097.1 843 Silent Mutation XP_016884586.1
XM_017029098.1 843 Silent Mutation XP_016884587.1
Gene
SEC14L6
Gene Name
SEC14 like lipid binding 6
There are no transcripts associated with this gene.

View Full Product Details