Product Details
- SNP ID
-
rs138445723
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:25235799 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCTGCTTTATGGAGTTTGACCT[C/T]GTAGTAATGGTCCTCACTTTGCTGA
- Phenotype
-
MIM: 602769
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNMT3A
PubMed Links
Gene Details
- Gene
- DNMT3A
- Gene Name
- DNA methyltransferase 3 alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320892.1 |
2706 |
Intron |
|
|
NP_001307821.1 |
NM_001320893.1 |
2706 |
Silent Mutation |
ACA,ACG |
T683T |
NP_001307822.1 |
NM_022552.4 |
2706 |
Silent Mutation |
ACA,ACG |
T835T |
NP_072046.2 |
NM_153759.3 |
2706 |
Silent Mutation |
ACA,ACG |
T646T |
NP_715640.2 |
NM_175629.2 |
2706 |
Silent Mutation |
ACA,ACG |
T835T |
NP_783328.1 |
NM_175630.1 |
2706 |
Intron |
|
|
NP_783329.1 |
XM_005264175.4 |
2706 |
Silent Mutation |
ACA,ACG |
T835T |
XP_005264232.1 |
XM_005264177.4 |
2706 |
Silent Mutation |
ACA,ACG |
T612T |
XP_005264234.1 |
XM_011532662.2 |
2706 |
Silent Mutation |
ACA,ACG |
T786T |
XP_011530964.1 |
XM_011532663.2 |
2706 |
Silent Mutation |
ACA,ACG |
T780T |
XP_011530965.1 |
XM_011532664.2 |
2706 |
Intron |
|
|
XP_011530966.1 |
XM_011532666.2 |
2706 |
Silent Mutation |
ACA,ACG |
T659T |
XP_011530968.1 |
XM_011532667.2 |
2706 |
Silent Mutation |
ACA,ACG |
T612T |
XP_011530969.1 |
XM_017003526.1 |
2706 |
Silent Mutation |
ACA,ACG |
T835T |
XP_016859015.1 |
XM_017003527.1 |
2706 |
Silent Mutation |
ACA,ACG |
T612T |
XP_016859016.1 |
View Full Product Details