Product Details

SNP ID
rs138604676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197706089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCACTCCAGCGGTGGCTGCGGGG[A/C]AACCCTCAGGCGATCACCCCCGAAC
Phenotype
MIM: 609728
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MARS2 PubMed Links

Gene Details

Gene
MARS2
Gene Name
methionyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138395.3 786 Silent Mutation GGA,GGC G228G NP_612404.1

View Full Product Details