Product Details

SNP ID
rs139138344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171316304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAACAAGTCCCCAAGTTGCTACT[G/T]AGCCAAACTGTAAAGGCCAGTCAGG
Phenotype
MIM: 609525
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
METTL8 PubMed Links

Gene Details

Gene
METTL8
Gene Name
methyltransferase like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321154.1 8972 UTR 3 NP_001308083.1
NM_001321155.1 8972 UTR 3 NP_001308084.1
NM_001321156.1 8972 UTR 3 NP_001308085.1
NM_001321157.1 8972 UTR 3 NP_001308086.1
NM_001321158.1 8972 UTR 3 NP_001308087.1
NM_001321159.1 8972 UTR 3 NP_001308088.1
NM_001321160.1 8972 UTR 3 NP_001308089.1
NM_001321161.1 8972 UTR 3 NP_001308090.1
NM_001321162.1 8972 UTR 3 NP_001308091.1
NM_024770.4 8972 UTR 3 NP_079046.2
XM_011511875.2 8972 Intron XP_011510177.1
XM_017004984.1 8972 Intron XP_016860473.1
XM_017004985.1 8972 Intron XP_016860474.1
XM_017004986.1 8972 Intron XP_016860475.1
XM_017004987.1 8972 Intron XP_016860476.1
XM_017004988.1 8972 Intron XP_016860477.1
XM_017004989.1 8972 Intron XP_016860478.1

View Full Product Details