Product Details

SNP ID
rs139229733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105241977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCAGGATCTCCTTGGCCATAG[C/T]GATGCTGCTGATGACCGTGGTGGGG
Phenotype
MIM: 604838 MIM: 606237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR45 PubMed Links

Gene Details

Gene
GPR45
Gene Name
G protein-coupled receptor 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007227.3 235 Missense Mutation GCG,GTG A40V NP_009158.3
Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
There are no transcripts associated with this gene.

View Full Product Details