Product Details
- SNP ID
-
rs139843084
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:73228508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCAAAGGTGGGGATGCTGGTGAC[A/G]TTGACTGGGATGGAAATGATGGCCC
- Phenotype
-
MIM: 605140
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCT7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11559241] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCT7
- Gene Name
- chaperonin containing TCP1 subunit 7
There are no transcripts associated with this gene.
- Gene
- PRADC1
- Gene Name
- protease associated domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032319.1 |
581 |
Silent Mutation |
AAC,AAT |
N171N |
NP_115695.1 |
- Gene
- SMYD5
- Gene Name
- SMYD family member 5
There are no transcripts associated with this gene.
View Full Product Details