Product Details

SNP ID
rs139901163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3192457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTGGTCACTGATGTCATCGT[C/T]GTCTACCAAGTGGCCGAAGGGCTCC
Phenotype
MIM: 608998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSSC1 PubMed Links

Gene Details

Gene
TSSC1
Gene Name
tumor suppressing subtransferable candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006711893.2 1044 Missense Mutation AAC,GAC N216D XP_006711956.1
XM_011510383.2 1044 Missense Mutation AAC,GAC N355D XP_011508685.1
XM_011510384.2 1044 Missense Mutation AAC,GAC N343D XP_011508686.1
XM_011510385.2 1044 Missense Mutation AAC,GAC N328D XP_011508687.1
XM_011510387.2 1044 Missense Mutation AAC,GAC N172D XP_011508689.1
XM_017004818.1 1044 Missense Mutation AAC,GAC N216D XP_016860307.1

View Full Product Details