Product Details

SNP ID
rs140034717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20203203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACTACAGCCGTATTCTCCCCC[A/G]AGGTTTCAAAGGTGAAGTCCTGTGG
Phenotype
MIM: 186355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDC1 PubMed Links

Gene Details

Gene
SDC1
Gene Name
syndecan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006946.1 1038 Missense Mutation TCG,TTG S216L NP_001006947.1
NM_002997.4 1038 Missense Mutation TCG,TTG S216L NP_002988.3
XM_005262620.4 1038 Missense Mutation TCG,TTG S240L XP_005262677.1
XM_005262621.3 1038 Missense Mutation TCG,TTG S211L XP_005262678.3
XM_005262622.2 1038 Missense Mutation TCG,TTG S198L XP_005262679.1

View Full Product Details