Product Details
- SNP ID
-
rs140090973
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:233034192 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAACCCATGCCCCTTGTATGACG[C/T]GCAGACGGGGACCCTCTTCCTCTTC
- Phenotype
-
MIM: 605528
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NEU2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2233388] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NEU2
- Gene Name
- neuraminidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005383.2 |
278 |
Missense Mutation |
GCG,GTG |
A93V |
NP_005374.2 |
View Full Product Details