Product Details

SNP ID
rs140632206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:182928814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAGGGCTGTACTGAGACATCAC[A/G]TTACTGGCCAGTGTTGGCAAAGAAA
Phenotype
MIM: 604891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCKAP1 PubMed Links

Gene Details

Gene
NCKAP1
Gene Name
NCK associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013436.4 3286 Silent Mutation AAC,AAT N1013N NP_038464.1
NM_205842.2 3286 Silent Mutation AAC,AAT N1019N NP_995314.1
XM_006712200.3 3286 Silent Mutation AAC,AAT N1017N XP_006712263.1
XM_006712201.3 3286 Silent Mutation AAC,AAT N1011N XP_006712264.1

View Full Product Details