Product Details
- SNP ID
-
rs140880883
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:20447526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGACGGCGCGTGTGGCAAGACGTGC[C/T]TGCTGATCGTGTTCAGTAAGGACGA
- Phenotype
-
MIM: 165370
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RHOB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7043] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RHOB
- Gene Name
- ras homolog family member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004040.3 |
456 |
Silent Mutation |
CTG,TTG |
L21L |
NP_004031.1 |
View Full Product Details