Product Details

SNP ID
rs141014675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3192427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGCCCTTACTTCTCTTCAGAAC[A/G]GTGGTCCTCCTGGTCACTGATGTCA
Phenotype
MIM: 608998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSSC1 PubMed Links

Gene Details

Gene
TSSC1
Gene Name
tumor suppressing subtransferable candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006711893.2 1074 Missense Mutation CGT,TGT R226C XP_006711956.1
XM_011510383.2 1074 Missense Mutation CGT,TGT R365C XP_011508685.1
XM_011510384.2 1074 Missense Mutation CGT,TGT R353C XP_011508686.1
XM_011510385.2 1074 Missense Mutation CGT,TGT R338C XP_011508687.1
XM_011510387.2 1074 Missense Mutation CGT,TGT R182C XP_011508689.1
XM_017004818.1 1074 Missense Mutation CGT,TGT R226C XP_016860307.1

View Full Product Details