Product Details
- SNP ID
-
rs141186317
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:74562382 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGAATTCTCTGGCTGTGTCCTGG[C/G]CCCGGTGGTTCCCCCTTGGCTAACA
- Phenotype
-
MIM: 602919
MIM: 607163
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DOK1
PubMed Links
Gene Details
- Gene
- DOK1
- Gene Name
- docking protein 1
There are no transcripts associated with this gene.
- Gene
- LOXL3
- Gene Name
- lysyl oxidase like 3
There are no transcripts associated with this gene.
- Gene
- M1AP
- Gene Name
- meiosis 1 associated protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001281295.1 |
402 |
Intron |
|
|
NP_001268224.1 |
NM_001281296.1 |
402 |
Silent Mutation |
GGC,GGG |
G372G |
NP_001268225.1 |
NM_001321739.1 |
402 |
Silent Mutation |
GGC,GGG |
G372G |
NP_001308668.1 |
NM_138804.4 |
402 |
Silent Mutation |
GGC,GGG |
G372G |
NP_620159.2 |
XM_005264152.3 |
402 |
Silent Mutation |
GGC,GGG |
G128G |
XP_005264209.1 |
XM_006711946.3 |
402 |
Silent Mutation |
GGC,GGG |
G410G |
XP_006712009.1 |
XM_011532548.2 |
402 |
Silent Mutation |
GGC,GGG |
G410G |
XP_011530850.1 |
XM_011532549.2 |
402 |
Silent Mutation |
GGC,GGG |
G390G |
XP_011530851.1 |
XM_011532550.2 |
402 |
Silent Mutation |
GGC,GGG |
G372G |
XP_011530852.1 |
XM_011532551.2 |
402 |
Silent Mutation |
GGC,GGG |
G372G |
XP_011530853.1 |
XM_011532552.2 |
402 |
Silent Mutation |
GGC,GGG |
G352G |
XP_011530854.1 |
XM_011532553.2 |
402 |
Intron |
|
|
XP_011530855.1 |
XM_011532554.2 |
402 |
Silent Mutation |
GGC,GGG |
G128G |
XP_011530856.1 |
View Full Product Details