Product Details

SNP ID
rs141478382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178469732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTCCGTGGTCCTTTGGTCACAG[A/C]ATAAAGTTCAATCTCAAAAATCAAT
Phenotype
MIM: 610219 MIM: 607062
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DFNB59 PubMed Links

Gene Details

Gene
DFNB59
Gene Name
deafness, autosomal recessive 59
There are no transcripts associated with this gene.

Gene
FKBP7
Gene Name
FK506 binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135212.1 556 Missense Mutation GCT,TCT A142S NP_001128684.1
NM_181342.2 556 Missense Mutation GCT,TCT A143S NP_851939.1
XM_005246638.4 556 Intron XP_005246695.1
XM_011511348.2 556 Missense Mutation GCT,TCT A36S XP_011509650.1
XM_011511349.2 556 Missense Mutation GCT,TCT A35S XP_011509651.1

View Full Product Details