Product Details

SNP ID
rs141630292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86815634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTTCAGGATCCATGGGTTAAG[A/C]AGCTTCTGTGAGGTTGTAGTATTGC
Phenotype
MIM: 186910 MIM: 186730
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD8A PubMed Links

Gene Details

Gene
CD8A
Gene Name
CD8a molecule
There are no transcripts associated with this gene.

Gene
CD8B
Gene Name
CD8b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178100.1 637 Missense Mutation TGC,TTC C193F NP_001171571.1
NM_004931.4 637 Intron NP_004922.1
NM_172101.3 637 UTR 3 NP_742099.1
NM_172102.3 637 Silent Mutation CTG,CTT L205L NP_742100.1
NM_172213.3 637 Silent Mutation CTG,CTT L235L NP_757362.1
XM_011533164.2 637 Intron XP_011531466.1

View Full Product Details