Product Details

SNP ID
rs142235643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219515052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCCGCTGCCCCAGCCCCAGTG[G/T]CGGGCTTCCCGGCTGTCACCCTCTG
Phenotype
MIM: 606715 MIM: 615495
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASIC4 PubMed Links

Gene Details

Gene
ASIC4
Gene Name
acid sensing ion channel subunit family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182847.2 883 Missense Mutation GCG,TCG A237S NP_878267.2
XM_005246671.2 883 Missense Mutation GCG,TCG A237S XP_005246728.2
XM_017004439.1 883 Intron XP_016859928.1
Gene
GMPPA
Gene Name
GDP-mannose pyrophosphorylase A
There are no transcripts associated with this gene.

View Full Product Details