Product Details

SNP ID
rs142826899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95148498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTTTCTCTTTGAGATGAATAC[C/T]GAAGCTTGAACTCTACTAATGGGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF514 PubMed Links

Gene Details

Gene
ZNF514
Gene Name
zinc finger protein 514
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318005.1 2808 UTR 3 NP_001304934.1
NM_032788.2 2808 UTR 3 NP_116177.1
XM_006712806.2 2808 UTR 3 XP_006712869.1

View Full Product Details