Product Details

SNP ID
rs143186655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119436898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACGGTTTTTTCCAGGCCCAGAGG[A/C]CTTTGGGCCAAAGGCAGCCCCGCCG
Phenotype
MIM: 182098
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCTR PubMed Links

Gene Details

Gene
SCTR
Gene Name
secretin receptor
There are no transcripts associated with this gene.

Gene
TMEM37
Gene Name
transmembrane protein 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183240.2 82 Missense Mutation ACT,CCT T11P NP_899063.2
XM_005263598.4 82 UTR 5 XP_005263655.1
XM_006712300.3 82 UTR 5 XP_006712363.1
XM_011510659.2 82 Missense Mutation ACT,CCT T23P XP_011508961.1

View Full Product Details