Product Details

SNP ID
rs143484996
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233713193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTCACCCTGGAGGTGAATATG[C/T]ACATCAAAGAAGAGAACTTTTTCAC
Phenotype
MIM: 606435 MIM: 606429 MIM: 606430 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
UGT1A10 PubMed Links
Additional Information
For this assay, SNP(s) [rs3755321] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 202 Intron NP_061948.1
Gene
UGT1A4
Gene Name
UDP glucuronosyltransferase family 1 member A4
There are no transcripts associated with this gene.

Gene
UGT1A5
Gene Name
UDP glucuronosyltransferase family 1 member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019078.1 202 Missense Mutation CAC,TAC H68Y NP_061951.1
Gene
UGT1A6
Gene Name
UDP glucuronosyltransferase family 1 member A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001072.3 202 Intron NP_001063.2
NM_205862.1 202 Intron NP_995584.1
Gene
UGT1A7
Gene Name
UDP glucuronosyltransferase family 1 member A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019077.2 202 Intron NP_061950.2
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 202 Intron NP_061949.3
Gene
UGT1A9
Gene Name
UDP glucuronosyltransferase family 1 member A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021027.2 202 Intron NP_066307.1

View Full Product Details