Product Details

SNP ID
rs143905411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202774848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAGCAAGAAAACCGGAAGTTT[A/G]AAACGTACACTACAAATGTAAAAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM117B PubMed Links

Gene Details

Gene
FAM117B
Gene Name
family with sequence similarity 117 member B
There are no transcripts associated with this gene.

Gene
ICA1L
Gene Name
islet cell autoantigen 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288622.1 6635 UTR 3 NP_001275551.1
NM_001288623.1 6635 UTR 3 NP_001275552.1
NM_001288624.1 6635 Intron NP_001275553.1
NM_138468.5 6635 UTR 3 NP_612477.3

View Full Product Details