Product Details

SNP ID
rs144640380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112832705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCTGGAGGTGGAGAGCTTTCAG[G/T]TCATATGGACCAGACATCACCAAGC
Phenotype
MIM: 147720
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL1B PubMed Links

Gene Details

Gene
IL1B
Gene Name
interleukin 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000576.2 510 Missense Mutation GAA,GAC E141D NP_000567.1
XM_017003988.1 510 Missense Mutation GAA,GAC E110D XP_016859477.1

View Full Product Details