Product Details

SNP ID
rs144948944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197569523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCTTCTAAAACAAGTTTTGAG[A/G]ATATGACAGACTAGTAAGTTTATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RFTN2 PubMed Links

Gene Details

Gene
RFTN2
Gene Name
raftlin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144629.2 4610 Intron NP_653230.2
XM_011510595.2 4610 UTR 3 XP_011508897.1
XM_011510596.2 4610 UTR 3 XP_011508898.1
XM_011510597.2 4610 UTR 3 XP_011508899.1
XM_011510598.2 4610 UTR 3 XP_011508900.1
XM_017003332.1 4610 UTR 3 XP_016858821.1

View Full Product Details