Product Details

SNP ID
rs145344229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219574980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGCTGCACCTGGCCACCTCTGC[C/T]CTCTCTCTGCTGACCCACCCCGTCC
Phenotype
MIM: 147380 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INHA PubMed Links

Gene Details

Gene
INHA
Gene Name
inhibin alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002191.3 698 Silent Mutation GCC,GCT A185A NP_002182.1
Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

View Full Product Details