Product Details
- SNP ID
-
rs145485033
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:20447474 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGGCCCGGCCCGCTCATGGCGGC[C/T]ATCCGCAAGAAGCTGGTGGTGGTGG
- Phenotype
-
MIM: 165370
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RHOB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7043] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RHOB
- Gene Name
- ras homolog family member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004040.3 |
404 |
Silent Mutation |
GCC,GCT |
A3A |
NP_004031.1 |
View Full Product Details