Product Details

SNP ID
rs145520463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99018171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTAGATACTGCTATTTGATCT[C/G]AAGAGAATGGATCCTTAAATAGACT
Phenotype
MIM: 607166
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSGA10 PubMed Links

Gene Details

Gene
TSGA10
Gene Name
testis specific 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025244.2 Intron NP_079520.1
NM_182911.3 Intron NP_878915.2
XM_005264023.1 Intron XP_005264080.1
XM_005264024.2 Intron XP_005264081.1
XM_005264025.1 Intron XP_005264082.1
XM_005264026.1 Intron XP_005264083.1
XM_005264027.1 Intron XP_005264084.1
XM_005264029.1 Intron XP_005264086.1
XM_006712781.2 Intron XP_006712844.1
XM_011511934.1 Intron XP_011510236.1
XM_011511935.2 Intron XP_011510237.1
XM_011511937.1 Intron XP_011510239.1
XM_011511939.1 Intron XP_011510241.1
XM_011511940.2 Intron XP_011510242.1
XM_011511941.1 Intron XP_011510243.1
XM_011511942.1 Intron XP_011510244.1
XM_011511943.2 Intron XP_011510245.1
XM_017005035.1 Intron XP_016860524.1
XM_017005036.1 Intron XP_016860525.1
XM_017005037.1 Intron XP_016860526.1
XM_017005038.1 Intron XP_016860527.1
XM_017005039.1 Intron XP_016860528.1
XM_017005040.1 Intron XP_016860529.1
XM_017005041.1 Intron XP_016860530.1
XM_017005042.1 Intron XP_016860531.1
XM_017005043.1 Intron XP_016860532.1
XM_017005044.1 Intron XP_016860533.1
XM_017005045.1 Intron XP_016860534.1
XM_017005046.1 Intron XP_016860535.1
XM_017005047.1 Intron XP_016860536.1
XM_017005048.1 Intron XP_016860537.1

View Full Product Details