Product Details
- SNP ID
-
rs145567470
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:201133101 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCATTAATTTTCCTCATGAAGGA[G/T]TACATGGGCCGAGGCAAGATAAGCA
- Phenotype
-
MIM: 603599
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CFLAR
PubMed Links
Gene Details
- Gene
- CFLAR
- Gene Name
- CASP8 and FADD like apoptosis regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001127183.2 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
NP_001120655.1 |
NM_001127184.2 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
NP_001120656.1 |
NM_001202515.1 |
762 |
Intron |
|
|
NP_001189444.1 |
NM_001202516.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
NP_001189445.1 |
NM_001202517.1 |
762 |
Missense Mutation |
GAG,GAT |
E22D |
NP_001189446.1 |
NM_001202518.1 |
762 |
Missense Mutation |
GAG,GAT |
E22D |
NP_001189447.1 |
NM_001202519.1 |
762 |
Missense Mutation |
GAG,GAT |
E22D |
NP_001189448.1 |
NM_001308042.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
NP_001294971.1 |
NM_001308043.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
NP_001294972.1 |
NM_003879.5 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
NP_003870.4 |
XM_017005189.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860678.1 |
XM_017005190.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860679.1 |
XM_017005191.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860680.1 |
XM_017005192.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860681.1 |
XM_017005193.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860682.1 |
XM_017005194.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860683.1 |
XM_017005195.1 |
762 |
Missense Mutation |
GAG,GAT |
E22D |
XP_016860684.1 |
XM_017005196.1 |
762 |
UTR 5 |
|
|
XP_016860685.1 |
XM_017005197.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860686.1 |
XM_017005198.1 |
762 |
Missense Mutation |
GAG,GAT |
E118D |
XP_016860687.1 |
- Gene
- CFLAR-AS1
- Gene Name
- CFLAR antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details