Product Details

SNP ID
rs145685280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108381789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACATGGACTCAGGAGATAGTGG[A/T]ATTAATACAAAATGAAGGTGATGTG
Phenotype
MIM: 608357
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SULT1C4 PubMed Links

Gene Details

Gene
SULT1C4
Gene Name
sulfotransferase family 1C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321770.1 624 Missense Mutation GAA,GTA E66V NP_001308699.1
NM_006588.3 624 Missense Mutation GAA,GTA E66V NP_006579.2
XM_017003807.1 624 UTR 5 XP_016859296.1

View Full Product Details