Product Details

SNP ID
rs146219183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112830456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATGTTTTCTGCTTGAGAGGTGC[A/T]GATGTACCAGTTGGGGAACTGGGCA
Phenotype
MIM: 147720
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL1B PubMed Links

Gene Details

Gene
IL1B
Gene Name
interleukin 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000576.2 802 Missense Mutation AGC,TGC S239C NP_000567.1
XM_017003988.1 802 Missense Mutation AGC,TGC S208C XP_016859477.1

View Full Product Details