Product Details

SNP ID
rs146264648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10046382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTGCATATGGATGCAGCCACAC[C/T]TGAACTACCAAAAGACTTCCATTCT
Phenotype
MIM: 603301
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLF11 PubMed Links

Gene Details

Gene
KLF11
Gene Name
Kruppel like factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177716.1 292 Missense Mutation CCT,CTT P75L NP_001171187.1
NM_001177718.1 292 Missense Mutation CCT,CTT P75L NP_001171189.1
NM_003597.4 292 Missense Mutation CCT,CTT P92L NP_003588.1
Gene
LOC101929882
Gene Name
uncharacterized LOC101929882
There are no transcripts associated with this gene.

View Full Product Details