Product Details

SNP ID
rs147001633
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.2:25234373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCAGTCTCTGCCTCGCCAAG[C/T]GGCTCATGTTGGAGACGTCAGTATA
Phenotype
MIM: 602769
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DNMT3A PubMed Links

Gene Details

Gene
DNMT3A
Gene Name
DNA methyltransferase 3 alpha
There are no transcripts associated with this gene.

View Full Product Details