Product Details

SNP ID
rs147488502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151251240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCATCCACTTTCTCCTGACTC[A/G]GAATGTGGAATTTTTTCAGTGTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM43 PubMed Links

Gene Details

Gene
RBM43
Gene Name
RNA binding motif protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198557.2 1345 Missense Mutation CCG,CTG P247L NP_940959.1
XM_017004052.1 1345 Missense Mutation CCG,CTG P250L XP_016859541.1

View Full Product Details