Product Details

SNP ID
rs147639954
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176152705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAAAGAATTTCATTTTAACAG[A/G]TATCTGACAAGGCGCCGTCGGATTG
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006898.4 1674 Intron NP_008829.3
XM_005246509.3 1674 Intron XP_005246566.1
XM_005246510.4 1674 Intron XP_005246567.1
XM_005246511.3 1674 Intron XP_005246568.1
XM_005246513.4 1674 Intron XP_005246570.1
XM_006712477.2 1674 Intron XP_006712540.1
XM_011511065.2 1674 Intron XP_011509367.1
XM_011511066.2 1674 Intron XP_011509368.1
Gene
HOXD4
Gene Name
homeobox D4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014621.2 1674 Silent Mutation AGA,AGG R177R NP_055436.2
XM_005246514.4 1674 Missense Mutation GAT,GGT D145G XP_005246571.1
Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details